Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MMP24
Entrez Gene ID
102176054
Gene Name
matrix metallopeptidase 24
Gene Aliases
-
Chromosome Location
Chr.13: 64062764 - 64116573 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018057758.1 XP_017913247.1 8 - 9 1562 92

View Full Product Details