Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TGFBR2
Entrez Gene ID
100861300
Gene Name
transforming growth factor beta receptor 2
Gene Aliases
TGF-BETARII
Chromosome Location
Chr.22: 5019183 - 5109133 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018067217.1 XP_017922706.1 4 - 5 1812 60
GenBank mRNA JO497463.2 - 3 - 4 534 60

View Full Product Details