Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TCTEX1D1
Entrez Gene ID
102178969
Gene Name
Tctex1 domain containing 1
Gene Aliases
-
Chromosome Location
Chr.3: 42350052 - 42378790 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005678296.3 XP_005678353.1 2 - 3 163 94

View Full Product Details