Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZFAND5
Entrez Gene ID
102171334
Gene Name
zinc finger AN1-type containing 5
Gene Aliases
-
Chromosome Location
Chr.8: 48393268 - 48404752 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018052069.1 XP_017907558.1 5 - 6 1406 92
XM_018052070.1 XP_017907559.1 5 - 6 628 92
GenBank mRNA JO428804.2 - 1 - 2 1702 92

View Full Product Details