Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MAD2L1BP
Entrez Gene ID
102176476
Gene Name
MAD2L1 binding protein
Gene Aliases
-
Chromosome Location
Chr.23: 31679212 - 31685279 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_013973797.2 XP_013829251.1 1 - 2 938 73

View Full Product Details