Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SPDYC
Entrez Gene ID
102171578
Gene Name
speedy/RINGO cell cycle regulator family member C
Gene Aliases
-
Chromosome Location
Chr.29: 43807625 - 43809170 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018042758.1 XP_017898247.1 2 - 3 205 75

View Full Product Details