Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PTGFRN
Entrez Gene ID
102178965
Gene Name
prostaglandin F2 receptor inhibitor
Gene Aliases
-
Chromosome Location
Chr.3: 94206114 - 94290611 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018045969.1 XP_017901458.1 7 - 8 3243 82

View Full Product Details