Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LAPTM4B
Entrez Gene ID
102180951
Gene Name
lysosomal protein transmembrane 4 beta
Gene Aliases
-
Chromosome Location
Chr.14: 15788487 - 15854655 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018058265.1 XP_017913754.1 6 - 7 830 81

View Full Product Details