Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KRT39
Entrez Gene ID
102178207
Gene Name
keratin 39
Gene Aliases
-
Chromosome Location
Chr.19: 40797168 - 40815645 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_013972066.2 XP_013827520.2 2 - 3 628 78
XM_013972067.2 XP_013827521.2 2 - 3 628 78
XM_013972068.2 XP_013827522.2 2 - 3 628 78
XM_013972069.2 XP_013827523.2 3 - 4 419 78
XM_018065050.1 XP_017920539.1 2 - 3 628 78

View Full Product Details