Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LRRC73
Entrez Gene ID
102179716
Gene Name
leucine rich repeat containing 73
Gene Aliases
GM88
Chromosome Location
Chr.23: 31783272 - 31786188 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_013973884.2 XP_013829338.1 5 - 6 979 70
XM_018039140.1 XP_017894629.1 4 - 5 840 70
GenBank mRNA JO644987.2 - 1 - 2 330 70

View Full Product Details