Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GCNA
Entrez Gene ID
102182112
Gene Name
germ cell nuclear acidic peptidase
Gene Aliases
ACRC
Chromosome Location
Chr.X|NW_017189516: 10276493 - 10301490 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_013976420.2 XP_013831874.1 8 - 9 3628 104
XM_018043749.1 XP_017899238.1 7 - 8 3718 104

View Full Product Details