Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MYNN
Entrez Gene ID
102181815
Gene Name
myoneurin
Gene Aliases
-
Chromosome Location
Chr.1: 97109422 - 97126961 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005675342.3 XP_005675399.1 2 - 3 393 93
XM_005675343.3 XP_005675400.1 2 - 3 393 93

View Full Product Details