Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OTOL1
Entrez Gene ID
102170553
Gene Name
otolin 1
Gene Aliases
-
Chromosome Location
Chr.1: 105425816 - 105440496 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018053789.1 XP_017909278.1 2 - 3 456 62

View Full Product Details