Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NT5DC4
Entrez Gene ID
102181400
Gene Name
5'-nucleotidase domain containing 4
Gene Aliases
-
Chromosome Location
Chr.11: 46073271 - 46081695 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018054714.1 XP_017910203.1 8 - 9 667 84

View Full Product Details