Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PCDH18
Entrez Gene ID
102188615
Gene Name
protocadherin 18
Gene Aliases
-
Chromosome Location
Chr.17: 50676342 - 50691114 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005691245.3 XP_005691302.1 2 - 3 2999 92
XM_005691246.3 XP_005691303.1 2 - 3 2996 92
GenBank mRNA JT709759.1 - 2 - 3 2985 92

View Full Product Details