Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
WDR76
Entrez Gene ID
102186346
Gene Name
WD repeat domain 76
Gene Aliases
-
Chromosome Location
Chr.21: 54945357 - 54994757 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_013973146.2 XP_013828600.1 2 - 3 673 68

View Full Product Details