Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GDF2
Entrez Gene ID
102178244
Gene Name
growth differentiation factor 2
Gene Aliases
-
Chromosome Location
Chr.28: 3482207 - 3486650 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005699334.3 XP_005699391.1 1 - 2 474 106
GenBank mRNA JO408691.2 - 1 - 2 295 106

View Full Product Details