Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HTR2C
Entrez Gene ID
102188025
Gene Name
5-hydroxytryptamine receptor 2C
Gene Aliases
-
Chromosome Location
Chr.X|NW_017189517: 29015430 - 29165761 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005700259.2 XP_005700316.1 3 - 4 611 85

View Full Product Details