Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ALG3
Entrez Gene ID
102181899
Gene Name
ALG3 alpha-1,3- mannosyltransferase
Gene Aliases
-
Chromosome Location
Chr.1: 82608064 - 82613536 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_013964097.2 XP_013819551.1 6 - 7 955 83
GenBank mRNA KA335133.1 - 3 - 4 543 83

View Full Product Details