Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PPP2R2C
Entrez Gene ID
102188298
Gene Name
protein phosphatase 2 regulatory subunit Bgamma
Gene Aliases
-
Chromosome Location
Chr.6: 103089844 - 103254694 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA JO631750.2 - 2 - 3 550 55
JO631751.2 - 2 - 3 550 55

View Full Product Details