Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LRRC24
Entrez Gene ID
102191631
Gene Name
leucine rich repeat containing 24
Gene Aliases
-
Chromosome Location
Chr.14: 81511375 - 81516339 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018058765.1 XP_017914254.1 1 - 2 1519 66
XM_018058766.1 XP_017914255.1 1 - 2 1519 66
XM_018058767.1 XP_017914256.1 2 - 3 488 66
XM_018058768.1 XP_017914257.1 2 - 3 355 66
XM_018058769.1 XP_017914258.1 2 - 3 379 66

View Full Product Details