Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ODF4
Entrez Gene ID
102171370
Gene Name
outer dense fiber of sperm tails 4
Gene Aliases
-
Chromosome Location
Chr.19: 27687843 - 27693366 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_013971973.2 XP_013827427.1 3 - 4 757 79

View Full Product Details