Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FADS1
Entrez Gene ID
102178247
Gene Name
fatty acid desaturase 1
Gene Aliases
-
Chromosome Location
Chr.29: 40597486 - 40611142 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018043052.1 XP_017898541.1 9 - 10 1118 75
XM_018043053.1 XP_017898542.1 9 - 10 1216 75
XM_018043054.1 XP_017898543.1 9 - 10 1125 75
XM_018043055.1 XP_017898544.1 9 - 10 998 75
GenBank mRNA JO623382.2 - 9 - 10 1237 75

View Full Product Details