Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZFP69
Entrez Gene ID
102185890
Gene Name
ZFP69 zinc finger protein
Gene Aliases
-
Chromosome Location
Chr.3: 15148976 - 15165364 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005678638.3 XP_005678695.2 5 - 6 1386 69
XM_005678639.3 XP_005678696.2 3 - 4 696 69
XM_018042342.1 XP_017897831.1 6 - 7 1072 69
XM_018042352.1 XP_017897841.1 6 - 7 1466 69

View Full Product Details