Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ETFBKMT
Entrez Gene ID
102183300
Gene Name
electron transfer flavoprotein subunit beta lysine methyltransferase
Gene Aliases
-
Chromosome Location
Chr.5: 76823263 - 76833423 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005680696.3 XP_005680753.1 2 - 3 697 91

View Full Product Details