Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FREM3
Entrez Gene ID
102190158
Gene Name
FRAS1 related extracellular matrix 3
Gene Aliases
-
Chromosome Location
Chr.17: 56643580 - 56743040 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005691247.2 XP_005691304.2 5 - 6 5871 66

View Full Product Details