Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NUTF2
Entrez Gene ID
102188247
Gene Name
nuclear transport factor 2
Gene Aliases
-
Chromosome Location
Chr.18: 36618243 - 36634560 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005692159.3 XP_005692216.1 2 - 3 557 101
XM_005692160.3 XP_005692217.1 2 - 3 244 101

View Full Product Details