Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BCLAF3
Entrez Gene ID
102186200
Gene Name
BCLAF1 and THRAP3 family member 3
Gene Aliases
CXHXorf23
Chromosome Location
Chr.X|NW_017189516: 50040370 - 50096850 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018043911.1 XP_017899400.1 4 - 5 960 95
XM_018043912.1 XP_017899401.1 3 - 4 683 95
XM_018043913.1 XP_017899402.1 3 - 4 807 95
XM_018043914.1 XP_017899403.1 4 - 5 960 95
XM_018043915.1 XP_017899404.1 4 - 5 960 95
XM_018043916.1 XP_017899405.1 4 - 5 960 95

View Full Product Details