Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FOLH1B
Entrez Gene ID
102190665
Gene Name
folate hydrolase 1B
Gene Aliases
-
Chromosome Location
Chr.29: 5342262 - 5414690 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018043532.1 XP_017899021.1 9 - 10 1226 100

View Full Product Details