Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RASSF10
Entrez Gene ID
102191354
Gene Name
Ras association domain family member 10
Gene Aliases
-
Chromosome Location
Chr.15: 42629410 - 42634529 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018059577.1 XP_017915066.1 1 - 1 1545 105
GenBank mRNA JT697918.1 - 1 - 1 454 105

View Full Product Details