Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DIPK1C
Entrez Gene ID
106503530
Gene Name
divergent protein kinase domain 1C
Gene Aliases
FAM69C
Chromosome Location
Chr.24: 4052615 - 4077564 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018039344.1 XP_017894833.1 4 - 4 1096 95

View Full Product Details