Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZDHHC22
Entrez Gene ID
102168834
Gene Name
zinc finger DHHC-type containing 22
Gene Aliases
-
Chromosome Location
Chr.10: 14740547 - 14754924 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018053924.1 XP_017909413.1 3 - 3 958 81
GenBank mRNA JO632557.2 - 3 - 3 956 81

View Full Product Details