The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_013965774.2 | XP_013821228.2 | 2 - 2 | 1826 | 114 |
GenBank mRNA | JO595712.2 | - | 2 - 2 | 749 | 114 |