Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gabarapl1
Entrez Gene ID
100135524
Gene Name
GABA type A receptor associated protein like 1
Gene Aliases
GEC-1, GEC1
Chromosome Location
Chr.Un|NT_176391: 4356662 - 4365125 on Build Cavpor3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001172950.1 NP_001166421.1 2 - 3 477 91
GenBank mRNA AF012920.2 - 2 - 3 477 91

View Full Product Details