Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cat
Entrez Gene ID
100135492
Gene Name
catalase
Gene Aliases
-
Chromosome Location
Chr.Un|NT_176358: 9620222 - 9620511 on Build Cavpor3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001172925.1 NP_001166396.1 4 - 5 515 103
GenBank mRNA AJ005111.1 - 4 - 5 515 103

View Full Product Details