Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nptx2
Entrez Gene ID
100135579
Gene Name
neuronal pentraxin 2
Gene Aliases
p50
Chromosome Location
Chr.Un|NT_176389: 6124012 - 6133686 on Build Cavpor3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001172728.1 NP_001166199.1 4 - 5 1187 84
GenBank mRNA U13234.1 - 4 - 5 1187 84
U13236.1 - 4 - 5 1195 84

View Full Product Details