Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sigmar1
Entrez Gene ID
100135589
Gene Name
sigma non-opioid intracellular receptor 1
Gene Aliases
Oprs1
Chromosome Location
Chr.Un|NT_176392: 15049803 - 15052333 on Build Cavpor3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001172992.1 NP_001166463.1 1 - 2 523 88
GenBank mRNA Z66537.1 - 1 - 2 523 88

View Full Product Details