Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tnnt2
Entrez Gene ID
100379561
Gene Name
troponin T2, cardiac type
Gene Aliases
-
Chromosome Location
Chr.Un|NT_176407: 21058748 - 21074196 on Build Cavpor3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001172863.1 NP_001166334.1 7 - 8 234 63
NM_001172864.1 NP_001166335.1 6 - 7 222 63
NM_001172865.1 NP_001166336.1 6 - 7 207 63
NM_001172866.1 NP_001166337.1 5 - 6 195 63
NM_001172867.1 NP_001166338.1 5 - 6 132 63
NM_001172868.1 NP_001166339.1 4 - 5 120 63
XM_005005810.2 XP_005005867.1 8 - 9 330 63
XM_013157812.1 XP_013013266.1 7 - 8 303 63
XM_013157813.1 XP_013013267.1 6 - 7 291 63
XM_013157814.1 XP_013013268.1 6 - 7 228 63
XM_013157815.1 XP_013013269.1 5 - 6 216 63
XM_013157816.1 XP_013013270.1 6 - 7 291 63
GenBank mRNA AF519741.1 - 7 - 8 234 63
AF519742.1 - 6 - 7 222 63
AF519743.2 - 6 - 7 207 63
AF519744.1 - 5 - 6 195 63
AF519745.1 - 5 - 6 132 63
AF519746.1 - 4 - 5 120 63

View Full Product Details