Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cryba2
Entrez Gene ID
100306966
Gene Name
crystallin beta A2
Gene Aliases
-
Chromosome Location
Chr.Un|NT_176330: 6184638 - 6187783 on Build Cavpor3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001173072.1 NP_001166543.1 3 - 4 344 74
GenBank mRNA EB371977.1 - 3 - 4 333 74
EF666995.1 - 3 - 4 344 74

View Full Product Details