Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cryab
Entrez Gene ID
100306972
Gene Name
crystallin alpha B
Gene Aliases
-
Chromosome Location
Chr.Un|NT_176400: 35864421 - 35869151 on Build Cavpor3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001173076.1 NP_001166547.1 1 - 2 236 99
GenBank mRNA EB371347.1 - 1 - 2 236 99
EU252027.1 - 1 - 2 236 99

View Full Product Details