Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Bfsp2
Entrez Gene ID
100306969
Gene Name
beaded filament structural protein 2
Gene Aliases
-
Chromosome Location
Chr.Un|NT_176411: 30599385 - 30662236 on Build Cavpor3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001173074.1 NP_001166545.1 5 - 6 1039 62
XM_005006539.2 XP_005006596.1 5 - 6 1021 62
GenBank mRNA EU257503.1 - 5 - 6 1039 62

View Full Product Details