The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001173095.1 | NP_001166566.1 | 4 - 5 | 803 | 73 |
GenBank mRNA | AF018569.1 | - | 4 - 5 | 803 | 73 |