Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cd1e
Entrez Gene ID
100379271
Gene Name
CD1e molecule
Gene Aliases
CD1-e
Chromosome Location
Chr.Un|NT_176363: 7686279 - 7689055 on Build Cavpor3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001172744.1 NP_001166215.1 2 - 3 365 79
GenBank mRNA AF145490.1 - 2 - 3 365 79

View Full Product Details