Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hrh4
Entrez Gene ID
100379272
Gene Name
histamine receptor H4
Gene Aliases
-
Chromosome Location
Chr.Un|NT_176405: 38525848 - 38538225 on Build Cavpor3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001172747.1 NP_001166218.1 2 - 3 456 88
GenBank mRNA AF358858.1 - 2 - 3 456 88

View Full Product Details