Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tacr1
Entrez Gene ID
100135626
Gene Name
tachykinin receptor 1
Gene Aliases
TAC1R
Chromosome Location
Chr.Un|NT_176382: 14010241 - 14160993 on Build Cavpor3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001172861.1 NP_001166332.1 2 - 3 622 63
NM_001172862.1 NP_001166333.1 2 - 3 605 63
GenBank mRNA AF428266.1 - 2 - 3 605 63
AF509591.1 - 2 - 3 605 63
X64323.1 - 2 - 3 622 63

View Full Product Details