Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pect
Entrez Gene ID
34716
Gene Name
Phosphoethanolamine cytidylyltransferase
Gene Aliases
CG5547, Dmel\CG5547, Dmel_CG5547, PECT, pect
Chromosome Location
Chr.2L: 13203762 - 13210828 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0032482
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_135769.3 NP_609613.2 6 - 7 1225 58
NM_165028.2 NP_723789.2 6 - 7 1291 58
NM_165029.2 NP_723790.2 6 - 7 1255 58
NM_165030.2 NP_723791.2 6 - 7 1234 58
GenBank mRNA BT003190.1 - 6 - 7 1249 58
BT125823.1 - 6 - 7 1002 58

View Full Product Details