Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- ZnT35C
- Entrez Gene ID
-
34890
- Gene Name
- Zinc transporter 35C
- Gene Aliases
-
BG:DS07295.1, CG3994, Dmel\CG3994, Dmel_CG3994, ZNT35C, cg3994, dZnT35C
- Chromosome Location
-
Chr.2L: 15228728 - 15247714 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0028516
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_135897.5 |
NP_609741.3 |
2 - 3 |
336 |
50 |
GenBank mRNA |
AY071460.1 |
- |
2 - 3 |
333 |
50 |
View Full Product Details