Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZnT35C
Entrez Gene ID
34890
Gene Name
Zinc transporter 35C
Gene Aliases
BG:DS07295.1, CG3994, Dmel\CG3994, Dmel_CG3994, ZNT35C, cg3994, dZnT35C
Chromosome Location
Chr.2L: 15228728 - 15247714 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0028516
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_135897.5 NP_609741.3 6 - 7 1247 79
NM_165105.3 NP_723901.1 5 - 6 1319 79
GenBank mRNA AY071460.1 - 6 - 7 1244 79

View Full Product Details