Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
jhamt
Entrez Gene ID
34977
Gene Name
juvenile hormone acid methyltransferase
Gene Aliases
BG:DS09218.5, CG17330, DmJHAMT, Dmel\CG17330, Dmel_CG17330, JAHMT, JHAMT, JHamt, Jhamt
Chromosome Location
Chr.2L: 16365903 - 16367039 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0028841
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001299051.1 NP_001285980.1 1 - 2 674 67
NM_135949.3 NP_609793.2 1 - 2 674 67

View Full Product Details