Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ChLD3
Entrez Gene ID
35002
Gene Name
CG17905-PA
Gene Aliases
CG17905, DmCDA3, DmChLD3, Dmel\CG17905, Dmel_CG17905
Chromosome Location
Chr.2L: 16738850 - 16742610 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0032598
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_135962.3 NP_609806.1 3 - 4 1600 85
GenBank mRNA AY118355.1 - 3 - 4 1598 85

View Full Product Details